EMAIL THIS PAGE TO A FRIEND
51590 Sigma-Aldrich

1-Heptadecanol

purum, ≥97.0% (GC)

Synonym: Heptadecyl alcohol

Purchase

Properties

Related Categories Alcohols, Biochemicals and Reagents, Building Blocks, C11 to C30+, Chemical Synthesis,
InChI Key   GOQYKNQRPGWPLP-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
mp   51-55 °C(lit.)
  51-55 °C

Price and Availability


Laboratory Gloves

The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
MI3885000
Flash Point(F) 
309.2 °F
Flash Point(C) 
154 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

241695 1-Heptadecanol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?