• USA Home
  • 51640 - Methyl heptadecanoate

EMAIL THIS PAGE TO A FRIEND
51640 Sigma-Aldrich

Methyl heptadecanoate

purum, ≥95.0% (GC)

Synonym: Heptadecanoic acid methyl ester, Methyl margarate

  • CAS Number 1731-92-6

  • Linear Formula CH3(CH2)15COOCH3

  • Molecular Weight 284.48

  •  Beilstein Registry Number 1782681

  •  EC Number 217-055-3

  •  MDL number MFCD00009001

  •  PubChem Substance ID 329757727

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis,
InChI Key   HUEBIMLTDXKIPR-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
bp   152-153 °C/0.05 mmHg(lit.)
mp   29.8-30.3 °C(lit.)

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

286087 Methyl heptadecanoate, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?