EMAIL THIS PAGE TO A FRIEND
51775 Sigma-Aldrich

3,5-Heptanedione

purum, ≥95.0% (GC)

  • CAS Number 7424-54-6

  • Linear Formula H3CCH2COCH2COCH2CH3

  • Molecular Weight 128.17

  •  Beilstein Registry Number 635979

  •  EC Number 231-054-5

  •  MDL number MFCD00015186

  •  PubChem Substance ID 329757740

Purchase

Properties

Related Categories Building Blocks, C7 to C8, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   DGCTVLNZTFDPDJ-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.455
  n20/D 1.456(lit.)
bp   175-177 °C/754 mmHg(lit.)
density   0.946 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN1224 - class 3 - PG 3 - Ketones, liquid, n.o.s., HI: all
WGK Germany 
3
Flash Point(F) 
134.6 °F
Flash Point(C) 
57 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

280070 3,5-Heptanedione, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?