EMAIL THIS PAGE TO A FRIEND
51790 Sigma-Aldrich

1-Heptanol

purum, ≥99.0% (GC)

Synonym: Heptyl alcohol

  • CAS Number 111-70-6

  • Linear Formula CH3(CH2)6OH

  • Molecular Weight 116.20

  •  Beilstein Registry Number 1731686

  •  EC Number 203-897-9

  •  MDL number MFCD00002986

Purchase

Properties

Related Categories Alcohols, Building Blocks, C7 to C8, Chemical Synthesis, Organic Building Blocks,
vapor pressure   0.5 mmHg ( 20 °C)
InChI Key   BBMCTIGTTCKYKF-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
autoignition temp.   662 °F
refractive index   n20/D 1.424(lit.)
  n20/D 1.424
bp   176 °C(lit.)
mp   −36 °C(lit.)
density   0.822 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
1
RTECS 
MK0350000
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

H2805 1-Heptanol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?