EMAIL THIS PAGE TO A FRIEND
51840 Sigma-Aldrich

1-Heptanethiol

purum, ≥98.0% (GC)

Synonym: Heptyl mercaptan, Mercaptan C7

  • CAS Number 1639-09-4

  • Linear Formula CH3(CH2)6SH

  • Molecular Weight 132.27

  •  Beilstein Registry Number 1731687

  •  EC Number 216-678-8

  •  FEMA Number 4259

  •  MDL number MFCD00004911

  •  PubChem Substance ID 329757753

Purchase

Properties

Related Categories Alkyl Thiols, Building Blocks, Chemical Synthesis, Materials Science, Micro/NanoElectronics,
InChI Key   VPIAKHNXCOTPAY-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.4520(lit.)
  n20/D 1.452
bp   173-176 °C/765 mmHg(lit.)
density   0.844 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3336PSN1A 3 / PGIII
WGK Germany 
3
RTECS 
MJ1400000
Flash Point(F) 
105.8 °F
Flash Point(C) 
41 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

H4506 1-Heptanethiol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?