EMAIL THIS PAGE TO A FRIEND
51856 Sigma-Aldrich

1-Heptene

analytical standard

  • CAS Number 592-76-7

  • Linear Formula CH3(CH2)4CH=CH2

  • Molecular Weight 98.19

  •  Beilstein Registry Number 1098332

  •  EC Number 209-767-8

  •  MDL number MFCD00009531

  •  PubChem Substance ID 329757755

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, Hydrocarbons,
grade   analytical standard
vapor pressure   101 mmHg ( 37.7 °C)
InChI Key   ZGEGCLOFRBLKSE-UHFFFAOYSA-N
assay   ≥99.5% (GC)
form   neat
autoignition temp.   505 °F
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.400(lit.)
  n20/D 1.400
bp   94 °C(lit.)
mp   −119 °C(lit.)
density   0.697 g/mL at 25 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2278 3 / PGII
WGK Germany 
2
RTECS 
MJ8840000
Flash Point(F) 
15.8 °F
Flash Point(C) 
-9 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

442267-U 1-Heptene, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?