EMAIL THIS PAGE TO A FRIEND
51930 Sigma-Aldrich

1-Hepten-3-ol

purum, ≥97.0% (GC)

Synonym: Butyl vinyl carbinol

  • CAS Number 4938-52-7

  • Linear Formula CH3(CH2)3CH(OH)CH=CH2

  • Molecular Weight 114.19

  •  Beilstein Registry Number 1720340

  •  EC Number 225-579-9

  •  MDL number MFCD00021940

  •  PubChem Substance ID 329757765

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
InChI Key   PZKFYTOLVRCMOA-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.433(lit.)
  n20/D 1.433
bp   153-156 °C(lit.)
density   0.836 g/mL at 20 °C
  0.836 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1987C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
129.2 °F
Flash Point(C) 
54 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?