• USA Home
  • 52037 - Herbimycin A from Streptomyces hygroscopicus

EMAIL THIS PAGE TO A FRIEND
52037 Sigma-Aldrich

Herbimycin A from Streptomyces hygroscopicus

≥98.0% (HPLC)

  • CAS Number 70563-58-5

  • Empirical Formula (Hill Notation) C30H42N2O9

  • Molecular Weight 574.66

  •  Beilstein Registry Number 4834067

  •  MDL number MFCD00151702

  •  PubChem Substance ID 329757778

Purchase

Properties

Related Categories Antibiotics, Antibiotics A to Z, Antibiotics G-M, Antibiotics by Application, Antineoplastic and Immunosuppressive Antibiotics,
InChI Key   MCAHMSDENAOJFZ-BVXDHVRPSA-N
assay   ≥98.0% (HPLC)
solubility   acetonitrile: soluble0.2 mg/mL, clear, yellow
Mode of action   enzyme | inhibits
antibiotic activity spectrum   viruses
storage temp.   −20°C

Description

Biochem/physiol Actions

An antibiotic that inhibits protein tyrosine kinase. Herbimycin A is a potent inhibitor of angiogenesis.

Other Notes

Inhibitor of protein-tyrosine kinases

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
LX8930000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

H6649 Herbimycin A from Streptomyces hygroscopicus, ≥95% (HPLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?