EMAIL THIS PAGE TO A FRIEND
52041 Sigma-Aldrich

Hexaethylene glycol

purum, ≥95.0% (GC)

Synonym: 3,6,9,12,15-Pentaoxaheptadecane-1,17-diol

  • CAS Number 2615-15-8

  • Linear Formula H(OCH2CH2)6OH

  • Molecular Weight 282.33

  •  Beilstein Registry Number 1638281

  •  EC Number 220-045-1

  •  MDL number MFCD00002877

  •  PubChem Substance ID 24874129

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
InChI Key   IIRDTKBZINWQAW-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.465(lit.)
  n20/D 1.465
bp   217 °C/4 mmHg(lit.)
mp   5-7 °C(lit.)
density   1.127 g/mL at 25 °C(lit.)

Description

Other Notes

Important starting material for preparing macrocyclic polyethers; building block in the synthesis of surfactants

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
MM3670000
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

259268 Hexaethylene glycol, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?