• USA Home
  • 52050 - Hexacyclen trisulfate salt

EMAIL THIS PAGE TO A FRIEND
52050 Sigma-Aldrich

Hexacyclen trisulfate salt

technical, ≥90% (CHN)

Synonym: 1,4,7,10,13,16-Hexaazacyclooctadecane trisulfate salt

  • CAS Number 56187-09-8

  • Empirical Formula (Hill Notation) C12H30N6 · 3H2SO4

  • Molecular Weight 552.64

  •  Beilstein Registry Number 5195914

  •  MDL number MFCD00013154

  •  PubChem Substance ID 329757781

Purchase

Properties

Related Categories Chelation/Complexation Compounds, Chemical Synthesis, Crown Ethers, Synthetic Reagents
InChI Key   UXVDKNLOZWKLJC-UHFFFAOYSA-N
grade   technical
assay   ≥90% (CHN)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?