EMAIL THIS PAGE TO A FRIEND
52090 Sigma-Aldrich

Hexachloroethane

technical, ≥95% (AT)

Synonym: Perchloroethane

  • CAS Number 67-72-1

  • Linear Formula Cl3CCCl3

  • Molecular Weight 236.74

  •  Beilstein Registry Number 1740341

  •  EC Number 200-666-4

  •  MDL number MFCD00000799

  •  PubChem Substance ID 329757786

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
vapor density   8.16 (vs air)
vapor pressure   0.4 mmHg ( 20 °C)
InChI Key   VHHHONWQHHHLTI-UHFFFAOYSA-N
grade   technical
assay   ≥95% (AT)
mp   183-185 °C (dec.)(lit.)
density   2.091 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?