EMAIL THIS PAGE TO A FRIEND
52191 Sigma-Aldrich

1-Hexacosanol

synthetic, ≥97.0% (GC)

Synonym: Ceryl alcohol, Hexacosyl alcohol

  • CAS Number 506-52-5

  • Linear Formula CH3(CH2)25OH

  • Molecular Weight 382.71

  •  Beilstein Registry Number 1783162

  •  EC Number 208-044-4

  •  MDL number MFCD00002940

  •  PubChem Substance ID 329757795

Purchase

Price and Availability


The Future of Bioimaging

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

H2139 1-Hexacosanol, ≥97% (capillary GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?