EMAIL THIS PAGE TO A FRIEND
52230 Sigma-Aldrich

Hexadecanedioic acid

purum, ≥98.0% (GC)

Synonym: Thapsic acid

  • CAS Number 505-54-4

  • Linear Formula HOOC(CH2)14COOH

  • Molecular Weight 286.41

  •  Beilstein Registry Number 1792831

  •  EC Number 208-013-5

  •  MDL number MFCD00002746

  •  PubChem Substance ID 329757804

Purchase

Properties

Related Categories Building Blocks, C13 to C42+, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   QQHJDPROMQRDLA-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
mp   120-123 °C(lit.)
  120-125 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

177504 Hexadecanedioic acid, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?