EMAIL THIS PAGE TO A FRIEND
52240 Sigma-Aldrich

Cetyl alcohol

puriss., ≥99.0% (GC)

Synonym: 1-Hexadecanol, Palmityl alcohol

  • CAS Number 36653-82-4

  • Linear Formula CH3(CH2)15OH

  • Molecular Weight 242.44

  •  Beilstein Registry Number 1748475

  •  EC Number 253-149-0

  •  MDL number MFCD00004760

Purchase

Properties

Related Categories Alcohols, Building Blocks, C11 to C30+, Chemical Synthesis, Organic Building Blocks,
InChI Key   BXWNKGSJHAJOGX-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
bp   190 °C/15 mmHg(lit.)
mp   49-50 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
MM0225000
Flash Point(F) 
338 °F
Flash Point(C) 
170 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

258741 1-Hexadecanol, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?