EMAIL THIS PAGE TO A FRIEND
52278 Sigma-Aldrich

1-Hexadecene

puriss., ≥99.0% (GC)

Synonym: Cetene

  • CAS Number 629-73-2

  • Linear Formula CH3(CH2)13CH=CH2

  • Molecular Weight 224.43

  •  Beilstein Registry Number 1753401

  •  EC Number 211-105-8

  •  MDL number MFCD00008991

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Carthamus tinctorius (Safflower oil), Cell Biology,
vapor density   >0.8 (vs air)
vapor pressure   <0.2 mmHg ( 20 °C)
InChI Key   GQEZCXVZFLOKMC-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
autoignition temp.   464 °F
refractive index   n20/D 1.441(lit.)
  n20/D 1.441
bp   274 °C(lit.)
mp   3-5 °C(lit.)
density   0.781 g/mL at 20 °C
  0.783 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
MM0479000
Flash Point(F) 
269.6 °F
Flash Point(C) 
132 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

H2131 1-Hexadecene, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?