• USA Home
  • 52349 - Hexadecylpyridinium chloride monohydrate

EMAIL THIS PAGE TO A FRIEND
52349 Sigma-Aldrich

Hexadecylpyridinium chloride monohydrate

≥98.0% (AT)

Synonym: Cetylpyridinium chloride monohydrate

  • CAS Number 6004-24-6

  • Empirical Formula (Hill Notation) C21H38ClN · H2O

  • Molecular Weight 358.00

  •  Beilstein Registry Number 3578606

  •  EC Number 204-593-9

  •  ISBN-10 MFCD00149977

  •  MDL number MFCD00149977

  •  PubChem Substance ID 329757822

Purchase

Description

Application

Used as a component of cationic and mixed micelles and in nanoparticle and hydrogel compositions and for extraction of membrane-associated enzymes such as alkaline phosphatase. Also used for extraction of chromium and palladium.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN2811 - class 6.1 - PG 2 - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
UU5075000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?