EMAIL THIS PAGE TO A FRIEND
523577 Sigma-Aldrich

Chromium(VI) oxide

crystalline

Synonym: Chromic anhydride

Purchase

Properties

Related Categories Chromium, Materials Science, Metal and Ceramic Science, Oxides
InChI Key   WGLPBDUCMAPZCE-UHFFFAOYSA-N
form   crystalline
mp   196 °C (dec.)(lit.)

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?