• USA Home
  • 52363 - Hexadecyltrimethylammonium phosphate monobasic concentrate

EMAIL THIS PAGE TO A FRIEND
52363 Sigma-Aldrich

Hexadecyltrimethylammonium phosphate monobasic concentrate

concentrate, for ion pair chromatography

Synonym: Cetyltrimethylammonium dihydrogen phosphate, Hexadecyltrimethylammonium dihydrogen phosphate concentrate

Purchase

Properties

Related Categories Acidic Ion Pair Reagents (Anionic), Analytical Reagents, Analytical/Chromatography, Buffers & Reagents, Chromatography Reagents & Solvents,
grade   for ion pair chromatography
InChI Key   ZSJUXFJODHVXFU-UHFFFAOYSA-M
quality   ampule
density   1.00 g/mL at 20 °C(lit.)
λ   in concentrate
UV absorption   λ: 210 nm Amax: 0.2
  λ: 230 nm Amax: 0.1
  λ: 290 nm Amax: 0.05
  λ: 330 nm Amax: 0.02
  λ: 500 nm Amax: 0.02
suitability   in accordance for filter test

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Packaging

concentrate in ampules for 6x1 l 0.005 M ready-to-use solution

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?