• USA Home
  • 52372 - Hexadecyltrimethylammonium chloride solution

EMAIL THIS PAGE TO A FRIEND
52372 Sigma-Aldrich

Hexadecyltrimethylammonium chloride solution

purum, ~25% in H2O

Synonym: Cetyltrimethylammonium chloride

  • CAS Number 112-02-7

  • Linear Formula CH3(CH2)15N(Cl)(CH3)3

  • Molecular Weight 320.00

  •  Beilstein Registry Number 3657974

  •  MDL number MFCD00011773

  •  PubChem Substance ID 329757831

Purchase

Properties

Related Categories Ammonium Salts, Biochemicals and Reagents, Cationic Detergents, Chemical Synthesis, Detergents,
description   cationic
InChI Key   WOWHHFRSBJGXCM-UHFFFAOYSA-M
grade   purum
concentration   ~25% in H2O
refractive index   n20/D 1.375(lit.)

Description

Other Notes

Preparation of the fairly stable permanganate salt which is a selective oxidizing agent

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

292737 Cetyltrimethylammonium chloride solution, 25 wt. % in H2O

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?