• USA Home
  • 52373 - Hexadecyltrimethylammonium bisulfate

EMAIL THIS PAGE TO A FRIEND
52373 Sigma-Aldrich

Hexadecyltrimethylammonium bisulfate

purum, ≥97.0% (T)

Synonym: Hexadecyltrimethylammonium hydrogen sulfate

  • CAS Number 68214-07-3

  • Linear Formula CH3(CH2)15N(HSO4)(CH3)3

  • Molecular Weight 381.61

  •  Beilstein Registry Number 8398798

  •  EC Number 269-286-4

  •  MDL number MFCD00134393

  •  PubChem Substance ID 24874365

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   UCRJJNVFJGKYQT-UHFFFAOYSA-M
grade   purum
assay   ≥97.0% (T)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

374598 Cetyltrimethylammonium hydrogensulfate, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?