• USA Home
  • 52512 - 1,1,1,3,3,3-Hexafluoro-2-propanol

EMAIL THIS PAGE TO A FRIEND
52512 Sigma-Aldrich

1,1,1,3,3,3-Hexafluoro-2-propanol

puriss., ≥99.0% (GC)

Synonym: HFP, Hexafluoroisopropanol

  • CAS Number 920-66-1

  • Linear Formula (CF3)2CHOH

  • Molecular Weight 168.04

  •  Beilstein Registry Number 1841007

  •  EC Number 213-059-4

  •  MDL number MFCD00011651

Purchase

Properties

Related Categories Alcohols, Building Blocks, C2 to C6, Chemical Synthesis, Organic Building Blocks,
InChI Key   BYEAHWXPCBROCE-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
impurities   ≤0.1% water
refractive index   n20/D 1.275(lit.)
bp   59 °C(lit.)
mp   −4 °C(lit.)
density   1.596 g/mL at 25 °C(lit.)

Description

Other Notes

Solvent with high solubilizing potential for peptides and peptide intermediates

This product has been replaced by 105228-ALDRICH | 1,1,1,3,3,3-Hexafluoro-2-propanol ≥99%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
2
RTECS 
UB6450000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?