EMAIL THIS PAGE TO A FRIEND
52516 Sigma-Aldrich

Hexafluoropropene

purum, ≥99.0%

Synonym: Hexafluoropropylene, Perfluoropropene

Purchase

Properties

Related Categories Chemical Synthesis, Specialty Gases, Synthetic Reagents More...
InChI Key   HCDGVLDPFQMKDK-UHFFFAOYSA-N
grade   purum
assay   ≥99.0%
bp   −28 °C(lit.)
mp   −153 °C(lit.)

Description

Packaging

Cylinder with net 50 g

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1858 2.2
WGK Germany 
1
RTECS 
UD0350000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

295388 Hexafluoropropene, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?