• USA Home
  • 52562 - (S)-(+)-4,4a,5,6,7,8-Hexahydro-4a-methyl-2(3H)-naphthalenone

EMAIL THIS PAGE TO A FRIEND
52562 Sigma-Aldrich

(S)-(+)-4,4a,5,6,7,8-Hexahydro-4a-methyl-2(3H)-naphthalenone

purum, ≥97.0% (sum of enantiomers, GC)

Synonym: (S)-(+)-10-Methyl-1(9)-octal-2-one, (S)-(+)-4a-Methyl-2,3,4,4a,5,6,7,8-octahydro-2-naphthalenone

  • CAS Number 4087-39-2

  • Empirical Formula (Hill Notation) C11H16O

  • Molecular Weight 164.24

  •  Beilstein Registry Number 2046096

  •  MDL number MFCD00064464

  •  PubChem Substance ID 329757854

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Ketones, Organic Building Blocks More...
InChI Key   OHERZLWVBJCXOF-NSHDSACASA-N
grade   purum
assay   ≥97.0% (sum of enantiomers, GC)
optical activity   [α]20/D +210±10°, neat
refractive index   n20/D 1.525
density   1.018 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?