52766 Sigma-Aldrich

Hexane

puriss., absolute, over molecular sieve (H2O ≤0.01%), ≥99.0% (GC)

DOWNLOAD MSDS (PDF)

Synonym: n-Hexane

  • CAS Number 110-54-3

  • Linear Formula CH3(CH2)4CH3

  • Molecular Weight 86.18

  •  Beilstein Registry Number 1730733

  •  EC Number 203-777-6

  •  MDL number MFCD00009520

  •  PubChem Substance ID 24874636

Properties

Related Categories Anhydrous Solvents, Solvent by Application, Solvents More...
vapor density   ~3 (vs air)
vapor pressure   256 mmHg ( 37.7 °C)
  5.2 psi ( 37.7 °C)
  ~132 mmHg ( 20 °C)
grade   absolute
  puriss.
assay   ≥99.0% (GC)
autoignition temp.   453 °F
quality   over molecular sieve (H2O ≤0.01%)
expl. lim.   7.7 %
refractive index   n20/D 1.375(lit.)
  n20/D 1.375
bp   69 °C(lit.)
mp   −95 °C(lit.)
density   0.659 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

Hexane
296090Hexane

anhydrous, 95%

Hexane
34859Hexane

CHROMASOLV®, for HPLC, ≥97.0% (GC)

Hexane
52767Hexane

for residue analysis, ≥99.0%

Hexane
52765Hexane

puriss. p.a., ACS reagent, ≥99.0% (GC)

Hexane
32293Hexane

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?