EMAIL THIS PAGE TO A FRIEND
52790 Sigma-Aldrich

Hexane Fraction

purum

Purchase

Properties

InChI Key   VLKZOEOYAKHREP-UHFFFAOYSA-N
grade   purum
refractive index   n20/D 1.38
bp   66-70 °C
density   0.67 g/mL at 20 °C

Description

Other Notes

Sales restrictions may apply

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Precautionary statements 
RIDADR 
UN1208 - class 3 - PG 2 - Hexanes
WGK Germany 
3
Flash Point(F) 
-14.8 °F
Flash Point(C) 
-26 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?