EMAIL THIS PAGE TO A FRIEND
52840 Sigma-Aldrich

1-Hexanol

purum, ≥98.0% (GC)

Synonym: Hexyl alcohol

  • CAS Number 111-27-3

  • Linear Formula CH3(CH2)5OH

  • Molecular Weight 102.17

  •  Beilstein Registry Number 969167

  •  EC Number 203-852-3

  •  MDL number MFCD00002982

Purchase

Properties

Related Categories Alcohols, Biochemicals and Reagents, Building Blocks, C2 to C6, Chemical Synthesis,
vapor density   4.5 (vs air)
vapor pressure   1 mmHg ( 25.6 °C)
InChI Key   ZSIAUFGUXNUGDI-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
autoignition temp.   559 °F
refractive index   n20/D 1.418(lit.)
  n20/D 1.418
bp   156-157 °C(lit.)
mp   −52 °C(lit.)
solubility   ethanol: soluble(lit.)
density   0.814 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2282 3 / PGIII
WGK Germany 
1
RTECS 
MQ4025000
Flash Point(F) 
140 °F
Flash Point(C) 
60 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?