• USA Home
  • 52863 - Sodium 1-hexanesulfonate monohydrate

EMAIL THIS PAGE TO A FRIEND
52863 Sigma-Aldrich

Sodium 1-hexanesulfonate monohydrate

≥99.0% (T)

Synonym: 1-Hexanesulfonic acid sodium salt monohydrate

  • CAS Number 207300-91-2

  • Linear Formula CH3(CH2)4CH2SO3Na · H2O

  • Molecular Weight 206.24

  •  Beilstein Registry Number 3727014

  •  EC Number 220-601-3

  •  MDL number MFCD00149549

  •  PubChem Substance ID 24877604

Purchase

Properties

Related Categories Alkyl Sulfonates, Anionic Detergents, Biochemicals and Reagents, Building Blocks, Chemical Synthesis,
description   anionic
InChI Key   CLCGFJYKZGFGSQ-UHFFFAOYSA-M
assay   ≥99.0% (T)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

52862 Sodium 1-hexanesulfonate monohydrate, for ion pair chromatography, ≥99.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?