• USA Home
  • 53020 - trans-3-Hexenedioic acid

EMAIL THIS PAGE TO A FRIEND
53020 Sigma-Aldrich

trans-3-Hexenedioic acid

purum, ≥99.0% (GC)

Synonym: 2-Butene-1,4-dicarboxylic acid, Dihydromuconic acid

  • CAS Number 4436-74-2

  • Linear Formula HO2CCH2CH=CHCH2CO2H

  • Molecular Weight 144.13

  •  Beilstein Registry Number 1723581

  •  EC Number 224-648-0

  •  MDL number MFCD00002785

  •  PubChem Substance ID 329757910

Purchase

Properties

InChI Key   YHGNXQAFNHCBTK-OWOJBTEDSA-N
grade   purum
assay   ≥99.0% (GC)
mp   195-196 °C(lit.)
  195-197 °C
solubility   methanol: soluble0.1 g/mL, clear

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?