EMAIL THIS PAGE TO A FRIEND
53131 Sigma-Aldrich

Hexylamine

purum, ≥98.0% (GC)

Synonym: 1-Aminohexane

  • CAS Number 111-26-2

  • Linear Formula CH3(CH2)5NH2

  • Molecular Weight 101.19

  •  Beilstein Registry Number 1731298

  •  EC Number 203-851-8

  •  MDL number MFCD00008240

  •  PubChem Substance ID 329757915

Purchase

Properties

InChI Key   BMVXCPBXGZKUPN-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
expl. lim.   2.1-9.3 %
refractive index   n20/D 1.418(lit.)
  n20/D 1.418
bp   131-132 °C(lit.)
mp   −23 °C(lit.)
density   0.766 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN2734 - class 8 - PG 1 - EHS - Amines, liquid, corrosive, flammable, n.o.s., HI: all
WGK Germany 
1
RTECS 
MQ4540000
Flash Point(F) 
80.6 °F
Flash Point(C) 
27 °C
Protocols & Articles

Articles

GC Analysis of Primary Amines on Carbowax® Amine

From our library of Articles, Sigma-Aldrich presents GC Analysis of Primary Amines on Carbowax® Amine
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

219703 Hexylamine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?