EMAIL THIS PAGE TO A FRIEND
53160 Sigma-Aldrich

Hexylbenzene

purum, ≥98.0% (GC)

Synonym: 1-Phenylhexane

Purchase

Properties

Related Categories Arenes, Building Blocks, Chemical Synthesis, Organic Building Blocks
InChI Key   LTEQMZWBSYACLV-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.486(lit.)
  n20/D 1.486
bp   226 °C(lit.)
mp   −61 °C(lit.)
density   0.861 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN3082 - class 9 - PG 3 - DOT NA1993 - Environmentally hazardous substances, liquid, n.o.s. HI: all (not BR)
WGK Germany 
3
Flash Point(F) 
181.4 °F
Flash Point(C) 
83 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P25701 Hexylbenzene, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?