• USA Home
  • 53302 - Histamine dihydrochloride

EMAIL THIS PAGE TO A FRIEND
53302 Sigma-Aldrich

Histamine dihydrochloride

tested according to Ph Eur

Synonym: 2-(4-Imidazolyl)ethylamine dihydrochloride, Histamini dihydrochloridum

  • CAS Number 56-92-8

  • Empirical Formula (Hill Notation) C5H9N3 · 2HCl

  • Molecular Weight 184.07

  •  Beilstein Registry Number 3624116

  •  EC Number 200-298-4

  •  MDL number MFCD00012703

  •  PubChem Substance ID 329757932

Purchase

Properties

Related Categories Amino Acids, Analytical/Chromatography, Building Blocks, C3 to C8, Chemical Synthesis,
InChI Key   PPZMYIBUHIPZOS-UHFFFAOYSA-N
assay   ≥98.5%
pharmacopeia   testing conforms to Pharmacopeia
mp   249-252 °C(lit.)

Description

Application

Histamine dihydrochloride was employed as pharmaceutical substance to investigate the orthogonality of the chromatographic systems, required for the separation of impurities from the active substance and from each other in drugs with an unknown impurity profile. It may be used in the preparation of imidazolium-linked cyclophane.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
MS1575000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?