EMAIL THIS PAGE TO A FRIEND
53321 Sigma-Aldrich

D-Histidine

puriss., ≥99.0% (T)

Synonym: (R)-2-Amino-3-(4-imidazolyl)propionic acid, D-α-Amino-β-(4-imidazolyl)propionic acid

  • CAS Number 351-50-8

  • Empirical Formula (Hill Notation) C6H9N3O2

  • Molecular Weight 155.15

  •  Beilstein Registry Number 84089

  •  EC Number 206-513-8

  •  MDL number MFCD00065963

  •  PubChem Substance ID 24877545

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Histidine, Peptide Chemistry More...
InChI Key   HNDVDQJCIGZPNO-RXMQYKEDSA-N
grade   puriss.
assay   ≥99.0% (T)
optical activity   [α]20/D +39±1°, c = 5% in H2O
mp   280-290 °C (dec.)
Gene Information   human ... CA1(759), CA2(760)

Description

Other Notes

Inactive isomer

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

H3751 D-Histidine, ≥98% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?