• USA Home
  • 53380 - D-Histidine monohydrochloride monohydrate

EMAIL THIS PAGE TO A FRIEND
53380 Sigma-Aldrich

D-Histidine monohydrochloride monohydrate

≥99.0% (AT)

Synonym: D-α-Amino-β-(4-imidazolyl)propionic acid monohydrochloride

  • CAS Number 6341-24-8

  • Empirical Formula (Hill Notation) C6H9N3O2 · HCl · H2O

  • Molecular Weight 209.63

  •  Beilstein Registry Number 4161356

  •  EC Number 228-733-3

  •  MDL number MFCD00150081

  •  PubChem Substance ID 329757874

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Histidine, Peptide Chemistry More...
InChI Key   CMXXUDSWGMGYLZ-ZJIMSODOSA-N
assay   ≥99.0% (AT)
optical activity   [α]20/D −10±0.5°, c = 5% in 5 M HCl
mp   ~180 °C (dec.)
  254 °C (dec.)(lit.)

Description

Other Notes

Unnatural isomer of histidine.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?