Synonym: Xylene mixture of isomers
CAS Number 1330-20-7
Linear Formula C6H4(CH3)2
Molecular Weight 106.17
MDL number MFCD00077264
PubChem Substance ID 24878011
Popular Documents: Specification Sheet (PDF)
| Related Categories | Amber Glass Bottles, Carbon Steel Cans with NPT Threads, Core Bioreagents, General Reagents, Histological, |
| grade | histological grade |
| vapor pressure | 18 mmHg ( 37.7 °C) |
| expl. lim. | 7 % |
| impurities | ≤25% ethylbenzene |
| evapn. residue | <0.002% |
| refractive index |
n |
| bp | 137-140 °C(lit.) |
20 L in steel drum
500 mL in glass bottle
4, 4×4 L in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?