EMAIL THIS PAGE TO A FRIEND
53540 Sigma-Aldrich

L-Homocystine

purum, ≥98.0% (NT)

Synonym: L-4,4′-Dithiobis(2-aminobutanoic acid)

  • CAS Number 626-72-2

  • Empirical Formula (Hill Notation) C8H16N2O4S2

  • Molecular Weight 268.35

  •  Beilstein Registry Number 1728583

  •  EC Number 210-962-5

  •  MDL number MFCD00020391

  •  PubChem Substance ID 24877552

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Homo-Amino Acids, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   ZTVZLYBCZNMWCF-WDSKDSINSA-N
grade   purum
assay   ≥98.0% (NT)
optical activity   [α]20/D +77±3°, c = 1% in 1 M HCl
mp   281-284 °C (dec.)(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?