EMAIL THIS PAGE TO A FRIEND
53585 Sigma-Aldrich

Homopiperazine

technical, ≥98.0% (NT)

Synonym: 1,4-Diazacycloheptane, 2,3,4,5,6,7-Hexahydro-1H-1,4-diazepine

  • CAS Number 505-66-8

  • Empirical Formula (Hill Notation) C5H12N2

  • Molecular Weight 100.16

  •  Beilstein Registry Number 102711

  •  EC Number 208-016-1

  •  MDL number MFCD00006933

  •  PubChem Substance ID 24878107

Purchase

Properties

Related Categories Building Blocks, C4 to C8, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
InChI Key   FQUYSHZXSKYCSY-UHFFFAOYSA-N
grade   technical
assay   ≥98.0% (NT)
bp   169 °C(lit.)
mp   38-40 °C(lit.)
  38-41 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3259PSN1 8 / PGII
WGK Germany 
1
RTECS 
HM3850000
Flash Point(F) 
141.8 °F
Flash Point(C) 
61 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?