EMAIL THIS PAGE TO A FRIEND
53630 Sigma-Aldrich

Homoveratronitrile

purum, ≥98.0% (GC)

Synonym: (3,4-Dimethoxyphenyl)acetonitrile, 3,4-Dimethoxybenzylcyanide

  • CAS Number 93-17-4

  • Linear Formula (CH3O)2C6H3CH2CN

  • Molecular Weight 177.20

  •  Beilstein Registry Number 1956100

  •  EC Number 202-225-1

  •  MDL number MFCD00001911

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Building Blocks, C10 to C27, Chemical Synthesis,
InChI Key   ASLSUMISAQDOOB-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
color   yellow
bp   171-178 °C/10 mmHg(lit.)
mp   62-63.5 °C(lit.)
  62-64 °C
solubility   methanol: soluble0.1 g/mL, clear

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
AL9325000
Flash Point(F) 
482 °F
Flash Point(C) 
250 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

126349 (3,4-Dimethoxyphenyl)acetonitrile, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?