• USA Home
  • 53790 - 2-Phenylpropionaldehyde

EMAIL THIS PAGE TO A FRIEND
53790 Sigma-Aldrich

2-Phenylpropionaldehyde

purum, ≥97.0% (GC)

Synonym: 2-Phenylpropanal, Hydratropaldehyde

  • CAS Number 93-53-8

  • Linear Formula CH3CH(C6H5)CHO

  • Molecular Weight 134.18

  •  Beilstein Registry Number 1905601

  •  EC Number 202-255-5

  •  MDL number MFCD00006973

  •  PubChem Substance ID 24878254

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C9, Carbonyl Compounds, Chemical Synthesis,
InChI Key   IQVAERDLDAZARL-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.517(lit.)
  n20/D 1.518
bp   92-94 °C/12 mmHg(lit.)
density   1.002 g/mL at 20 °C
  1.002 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
CY1460000
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?