• USA Home
  • 53855 - Hydrazine hydrate solution

EMAIL THIS PAGE TO A FRIEND
53855 Sigma-Aldrich

Hydrazine hydrate solution

purum, ~25% in H2O

Synonym: Hydrazinium hydroxide

  • CAS Number 10217-52-4

  • Linear Formula NH2NH2 · H2O

  • Molecular Weight 50.06

  •  Beilstein Registry Number 878137

  •  MDL number MFCD00149931

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Hydrazines, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   IKDUDTNKRLTJSI-UHFFFAOYSA-N
grade   purum
concentration   ~25% in H2O
density   1.03 g/mL at 20 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN 3293 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?