• USA Home
  • 54102 - Hydrocortisone 21-acetate

EMAIL THIS PAGE TO A FRIEND
54102 Sigma-Aldrich

Hydrocortisone 21-acetate

tested according to Ph.Eur.

Synonym: 11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 21-acetate, 17-α-Hydroxycorticosterone acetate, 17-Hydroxycorticosterone 21-acetate, 21-Acetoxy-4-pregnene-11β,17α-diol-3,20-dione, 4-Pregnene-11β,17α,21-triol-3,20-dione 21-acetate, Hydrocortisoni acetas

  • CAS Number 50-03-3

  • Empirical Formula (Hill Notation) C23H32O6

  • Molecular Weight 404.50

  •  Beilstein Registry Number 2066841

  •  EC Number 200-004-4

  •  MDL number MFCD00037714

  •  PubChem Substance ID 24878508

Purchase

Properties

Related Categories Antibiotics, Antibiotics A to Z, Antibiotics G-M, Biochemicals and Reagents, Core Bioreagents,
InChI Key   ALEXXDVDDISNDU-JZYPGELDSA-N
pharmacopeia   testing conforms to Pharmacopeia
mp   223 °C (dec.)(lit.)
Gene Information   human ... SERPINA6(866)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Hydrocortisone 21-acetate is used as a stabilized form of the glucocorticoid in studies on stress and homeostasis.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
GM8960000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?