EMAIL THIS PAGE TO A FRIEND
54191 Sigma-Aldrich

1-Adamantanol

purum, ≥99.0% (GC)

Synonym: 1-Hydroxyadamantane

  • CAS Number 768-95-6

  • Empirical Formula (Hill Notation) C10H16O

  • Molecular Weight 152.23

  •  Beilstein Registry Number 1903952

  •  EC Number 212-202-8

  •  MDL number MFCD00074729

  •  PubChem Substance ID 24878568

Purchase

Properties

Related Categories Alcohols, Building Blocks, C9 to C10, Chemical Synthesis, Organic Building Blocks,
InChI Key   VLLNJDMHDJRNFK-CHIWXEEVSA-N
grade   purum
assay   ≥99.0% (GC)
mp   247 °C (subl.)(lit.)
  265-290 °C (desc.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
AU4980000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

130346 1-Adamantanol, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?