EMAIL THIS PAGE TO A FRIEND
54464 Sigma-Aldrich

HEPES potassium salt

≥97.0% (dried material, NT)

Synonym: 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid potassium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) potassium salt

  • CAS Number 82207-62-3

  • Empirical Formula (Hill Notation) C8H17KN2O4S

  • Molecular Weight 276.39

  •  MDL number MFCD00043438

Purchase

Properties

InChI Key   KWURIFKZTJGMFD-UHFFFAOYSA-M
assay   ≥97.0% (dried material, NT)
impurities   ≤1% water
useful pH range   6.8 - 8.2
pKa    7.5
solubility   H2O: soluble50 mg/mL, clear, colorless

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

H0527 HEPES potassium salt, ≥99.5% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?