• USA Home
  • 54518 - 19-Hydroxy-4-androstene-3,17-dione

EMAIL THIS PAGE TO A FRIEND
54518 Sigma-Aldrich

19-Hydroxy-4-androstene-3,17-dione

technical

Synonym: 4-Androsten-19-ol-3,17-dione, 4-Androstene-3,17-dione-19-ol

  • CAS Number 510-64-5

  • Empirical Formula (Hill Notation) C19H26O3

  • Molecular Weight 302.41

  •  Beilstein Registry Number 2567249

  •  EC Number 208-116-5

  •  MDL number MFCD00003616

  •  PubChem Substance ID 329758034

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Complex Molecules
InChI Key   XGUHPTGEXRHMQQ-BGJMDTOESA-N
grade   technical
optical activity   [α]20/D +190±5°, c = 1% in chloroform

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?