• USA Home
  • 54600 - 4-Hydroxybenzhydrazide

EMAIL THIS PAGE TO A FRIEND
54600 Sigma-Aldrich

4-Hydroxybenzhydrazide

suitable for fluorescence, ≥97.0% (T)

Synonym: 4-Hydroxybenzoylhydrazine, 4-Hydroxybenzoic hydrazide, p-Hydroxybenzoic acid hydrazide

  • CAS Number 5351-23-5

  • Linear Formula HOC6H4CONHNH2

  • Molecular Weight 152.15

  •  Beilstein Registry Number 473052

  •  EC Number 226-326-5

  •  MDL number MFCD00007605

Purchase

Properties

Related Categories Biochemicals and Reagents, Detection, Fluorescent Indicators and Probes, Fluorescent Labels, Fluorescent Probes, Labels, Particles and Stains,
InChI Key   ZMZGIVVRBMFZSG-UHFFFAOYSA-N
assay   ≥97.0% (T)
mp   264-266 °C (dec.)(lit.)
fluorescence   λex 360 nm; λem 425 nm (after reaction with 5-hydroxymethyl-2-furaldehydezinc acetate)
suitability   suitable for fluorescence

Description

Other Notes

Reagent for the rapid automated determination of serum glucose

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

H9882 4-Hydroxybenzhydrazide, ≥97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?