• USA Home
  • 54840 - 2-Hydroxybenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
54840 Sigma-Aldrich

2-Hydroxybenzyl alcohol

purum, ≥98.0% (HPLC)

Synonym: Salicyl alcohol, Saligenin

Purchase

Properties

Related Categories Alcohols, Building Blocks, C7 to C8, Chemical Synthesis, Organic Building Blocks,
InChI Key   CQRYARSYNCAZFO-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   83-85 °C(lit.)
  84-86 °C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DO6430000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?