• USA Home
  • 55000 - Ethyl 2-hydroxyhexanoate

EMAIL THIS PAGE TO A FRIEND
55000 Sigma-Aldrich

Ethyl 2-hydroxyhexanoate

purum, ≥98.0% (GC)

Synonym: Ethyl 2-hydroxycaproate

  • CAS Number 52089-55-1

  • Linear Formula CH3(CH2)3CH(OH)COOC2H5

  • Molecular Weight 160.21

  •  Beilstein Registry Number 1704446

  •  EC Number 257-655-2

  •  MDL number MFCD00004583

  •  PubChem Substance ID 24879083

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   MRYSSTRVUMCKKB-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.424(lit.)
  n20/D 1.425
bp   195 °C(lit.)
density   0.967 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
208.4 °F
Flash Point(C) 
98 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

232580 Ethyl 2-hydroxyhexanoate, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?