EMAIL THIS PAGE TO A FRIEND
55005 Sigma-Aldrich

N-Boc-hydroxylamine

purum, ≥98.0% (N)

Synonym: tert-Butyl N-hydroxycarbamate

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Protected Amines More...
InChI Key   DRDVJQOGFWAVLH-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (N)
mp   53-55 °C(lit.)
  53-55 °C
storage temp.   2-8°C

Description

Other Notes

Reagent for the synthesis of α-aminoxy acids ; intermediate for the preparation of hydroxamic acids ; mild oxidation of hydroxamic acids to acyl nitroso compds. and CA

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?