EMAIL THIS PAGE TO A FRIEND
55067 Sigma-Aldrich

6-Quinolinol

technical, ≥95% (HPLC)

Synonym: 6-Hydroxyquinoline

  • CAS Number 580-16-5

  • Empirical Formula (Hill Notation) C9H7NO

  • Molecular Weight 145.16

  •  Beilstein Registry Number 113196

  •  EC Number 209-454-6

  •  MDL number MFCD00047611

  •  PubChem Substance ID 329758073

Purchase

Properties

Related Categories Building Blocks, C8 to C10, Chemical Synthesis, Heterocyclic Building Blocks, Quinolines More...
InChI Key   OVYWMEWYEJLIER-UHFFFAOYSA-N
grade   technical
assay   ≥95% (HPLC)
mp   188-190 °C(lit.)
  192-196 °C

Description

Application

6-Quinolinol may be used in the preparation of the following heterocyclic compounds:
• 1,2-dihydro-1-aryl[1,3]oxazino[5,6-f]quinolin-3-ones
• highly substituted 4H-chromene derivatives
• dioxino(5,4-f)quinoline
• methyl 3-oxo-3H-pyrano[3,2-f]quinoline-1-carboxylate

General description

6-Quinolinol can be prepared by refluxing a mixture of 6-methoxyquinoline bisulfate and concentrated hydrobromic acid followed by neutralization with ammonia.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
VC4130000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?