• USA Home
  • 55110 - 8-Hydroxy-5-quinolinesulfonic acid monohydrate

EMAIL THIS PAGE TO A FRIEND
55110 Sigma-Aldrich

8-Hydroxy-5-quinolinesulfonic acid monohydrate

technical, ≥95% (T)

  • CAS Number 283158-18-9

  • Empirical Formula (Hill Notation) C9H7NO4S · H2O

  • Molecular Weight 243.24

  •  Beilstein Registry Number 195247

  •  EC Number 201-570-5

  •  MDL number MFCD00149482

  •  PubChem Substance ID 24879197

Purchase

Properties

Related Categories Building Blocks, C8 to C10, Chemical Synthesis, Heterocyclic Building Blocks, Quinolines More...
InChI Key   WUXYGIQVWKDVTJ-UHFFFAOYSA-N
grade   technical
assay   ≥95% (T)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

H58757 8-Hydroxy-5-quinolinesulfonic acid hydrate, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?