• USA Home
  • 55275 - Dimethyl 3-hydroxyglutarate

EMAIL THIS PAGE TO A FRIEND
55275 Sigma-Aldrich

Dimethyl 3-hydroxyglutarate

purum, ≥97.0% (GC)

  • CAS Number 7250-55-7

  • Linear Formula CH3OCOCH2CH(OH)CH2COOCH3

  • Molecular Weight 176.17

  •  Beilstein Registry Number 1776666

  •  MDL number MFCD00042841

  •  PubChem Substance ID 329758093

Purchase

Properties

Related Categories Building Blocks, C6 to C7, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   CUPGMRSSZADEIW-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.442(lit.)
  n20/D 1.445
bp   139-140 °C/8 mmHg(lit.)
density   1.192 g/mL at 25 °C(lit.)

Description

Other Notes

Starting material for the synthesis of pimaricin

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

166979 Dimethyl 3-hydroxyglutarate, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?